Learn More
Medchemexpress LLC Mepivacaine | 96-88-8 | 98.10% | 1 ML
Supplier: Medchemexpress LLC HYB051710MM/1ML
Mepivacaine is an amide-type local anesthetic agent that binds to specific voltage-gated sodium ion channels in neuronal cell membranes, inhibiting both sodium influx and membrane depolarization, leading to a blockage of nerve impulse initiation and conduction, and a reversible loss of sensation.
- Binds to specific voltage-gated sodium ion channels
- Inhibits sodium influx and membrane depolarization
- Blocks nerve impulse initiation and conduction
- Provides reversible loss of sensation
- Has a rapid onset and moderate duration of action