Learn More
Medchemexpress LLC Meptyldinocap | 131-72-6 | 99.9% | 364.39 | 1 ML

Supplier: Medchemexpress LLC HY1752210MM/1ML
Meptyldinocap (2,4-DNOPC) is a novel fungicide effective against powdery mildew (Erysiphe necator). It exhibits both protectant and post-infective activities and is intended for research use only.
- High purity of 99.91%.
- Effective against powdery mildew (Erysiphe necator).
- Exhibits protectant and post-infective activities.
- Soluble in DMSO (≥ 150 mg/mL) and ethanol (100 mg/mL).
- Stable for extended periods when stored properly: 3 years at -20°C in pure form.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.