Learn More
Medchemexpress LLC Mequitazine | 29216-28-2 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB216810MM/1ML
Encompass_Preferred
Mequitazine is a potent, and long-acting histamine H1 antagonist. It is a potent H1-receptors selective antihistaminic drug widely studied and used for allergic disorders such as hay fever and urticaria.
  • Potent, and long-acting histamine H1 antagonist.
  • Potent H1-receptors selective antihistaminic drug.
  • Widely studied and used for allergic disorders such as hay fever and urticaria.
  • Demonstrates significant bactericidal effects against all the tested clinical isolates including Ps. aeruginosa.
  • Its effect against the Gram-positive isolates is more pronounced.
Catalog No. 50-004-12275