Learn More
Cayman Chemical Mer-NF5003F 1mg
SDP
Supplier:  Cayman Chemical 299611 MG
Encompass_Preferred
A sesquiterpene with diverse biological activities; inhibits AMV protease (IC50 = 7.8 µM); inhibits ST6N , ST3O, and ST3N (IC50s = 0.61, 6.7, and 10 UG/ml, respectively), as well as fucosyltransferase (IC50 = 11.3 UG/ml); active against HSV-1 in vitro (IC50 = 4.32 UG/ml); active against the multidrug-resistant P. falciparum strain K1 (IC50 = 0.85 UG/ml)
Catalog No. 50-298-8548