Learn More
Selleck Chemical LLC Merbromin
SDP
Supplier:  Selleck Chemical LLC S6872500MG
Encompass_Preferred
Merbromin (Mercurochrome) a xanthene dye and an antibacterial agent is the first of a series of antiseptics that contained mercury also is a mixed-type inhibitor of 3CLpro due to its ability of increasing the KM and decreasing the Kcat of 3CLpro
Catalog No. 50-000-02633