Learn More
Crescent Chemical Co Inc MERCAPTOETHANOL MOL BIO GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE39563.01
Encompass_Preferred
Mercaptoethanol molecular biology grade DNase/RNase not detected. Size - 50ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 39563.01 CAS 60-24-2
Catalog No. 50-248-390