Learn More
Gelest Inc [4-6% (mercaptopropyl)methylsiloxane] -Dimethylsiloxane copolymer, 120-170 cSt | 102783-03-9 | MFCD00239549 | 100g
Supplier:  Gelest Inc SMS042100G
Encompass
This item has a minimum qty of 14 per supplier requirements.
[4-6% (mercaptopropyl)methylsiloxane] -Dimethylsiloxane copolymer, 120-170 cSt | MW: 6,000-8,000 | 102783-03-9 | MFCD00239549 | 100g
Catalog No. NC1391066