Learn More
Hach Company Mercuric Nitrate Digital Titrator Cartridge, 2.570 N
Supplier:  Hach Company 2393701
Encompass
For determination of Salinity by Digital Titrator titration. 2.570 N. Digits x 0.1 = ppt Salinity in 2 mL sample.
Catalog No. 50-199-5908