Learn More
ERA WATERS CORP MERCURY, WS
SDP
Supplier:  ERA WATERS CORP 551
Encompass
One 15 mL screw-cap vial yields up to 1 liter after dilution. Use with CVAA, ICP-MS or CVAFS methods.
Catalog No. NC9695162