Learn More
Beaver Visitec Intl Ltd MEROCEL SPONGE POINTS, 400 COUNT, 20/POUCH
Supplier:  Beaver Visitec Intl Ltd 400115
Encompass
Made from exclusive Merocel polyvinyl acetal (PVA)material. The Merocel material is highly absorbent, fastwicking and lint free. 400 count, 20 per pouch,20 pouches per box
Catalog No. NC0930579