Learn More
TARGETMOL CHEMICALS INC MERODANTOIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T332965MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Merodantoin is an apoptosis inducer and triggers cell apoptosis associated with ROS-dependent autophagy. Merodantoin targets cancer cells expressing KRAS mutations involved in Akt-dependent ROS production. purity: 100%
Catalog No. 50-258-0314