Learn More
Medchemexpress LLC Meropenem-d6 | 1217976-95-8 | MFCD13195528 | 98.0% | 389.50 g/mol | C17H19D6N3O5S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13678S5MG
Encompass_Preferred
Meropenem-d6 is the deuterium-labeled analogue of the carbapenem antibiotic meropenem, provided as a solid analytical standard for use in mass spectrometry. It is intended for research use as an internal standard, quality control material, and for method development and validation in LC-MS/MS workflows.
  • Deuterium-labeled internal standard for quantitative LC-MS/MS.
  • High purity suitable for analytical workflows.
  • Provided in milligram-scale vials for small-batch experiments.
  • Supports method development, validation, and quality control.
  • Stable isotope labeling enables reliable mass shift for accurate quantitation.
Catalog No. 50-002-21855