Learn More
Medchemexpress LLC Meropenem (trihydrate) | 119478-56-7 | 99.9% | 437.51 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY13678A10MM/1ML
Encompass_Preferred
Meropenem trihydrate is a carbapenem antibiotic with broad-spectrum antibacterial activity. It is intrinsically stable to dehydropeptidase-1 (DHP-1) degradation and acts by inhibiting bacterial cell wall synthesis by binding to and inactivating penicillin-binding proteins (PBPs).
  • Active against susceptible and resistant *N. gonorrhoeae* (MIC value of 0.02-0.06 mg/mL)
  • Active against *H. influenzae* (MIC value of 0.03-0.12 mg/mL)
  • Active against *H. ducreyi* (MIC value of 0.015-0.12 mg/mL)
  • Possesses broad-spectrum *in vitro* activity against many Gram-positive, Gram-negative, and anaerobic bacteria
  • Lacks activity against *Enterococcus faecium*
  • Lacks activity against methicillin-resistant *Staphylococcus aureus*
  • Lacks activity against *Stenotrophomonas maltophilia*
Catalog No. 50-003-51708