Learn More
Medchemexpress LLC Meropenem trihydrate | 119478-56-7 | 99.9% | 437.51 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY13678A
Encompass_Preferred
Meropenem trihydrate is a carbapenem antibiotic with broad-spectrum antibacterial activity. It inhibits bacterial cell wall synthesis by binding to and inactivating penicillin-binding proteins (PBPs).
  • Intrinsically stable to dehydropeptidase-1 (DHP-1) degradation
  • Broad-spectrum in vitro activity against many Gram-positive, Gram-negative, and anaerobic bacteria
  • Targets β-lactam
  • Active against susceptible and resistant N. gonorrhoeae, H. influenzae, and H. ducreyi
Catalog No. 50-003-47946