Learn More
Medchemexpress LLC Meropenem trihydrate | 119478-56-7 | MFCD08600005 | 100.0% | 437.51 g/mol | C17H31N3O8S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY13678AR5MG
Encompass_Preferred
Meropenem trihydrate is the analytical standard form of the carbapenem antibiotic meropenem, provided as a high-purity reference material for research, method development, and quality control applications. The trihydrate form ensures a defined hydration state for consistent analytical results.
  • High purity suitable for analytical and QC applications.
  • Molecular weight 437.51 g/mol for accurate calculations.
  • Provided as a 5 mg pack for small-scale testing.
  • Trihydrate form ensures consistent hydration state.
  • Suitable for method development, calibration, and potency testing.
  • Stable when stored under recommended conditions for reference standards.
Catalog No. 50-002-21956