Learn More
Medchemexpress LLC Meropenem trihydrate | 119478-56-7 | MFCD08600005 | 99.9% | 437.51 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY13678A200MG
Encompass_Preferred
Meropenem trihydrate is a carbapenem antibiotic with broad-spectrum antibacterial activity. It inhibits bacterial cell wall synthesis by binding to and inactivating penicillin-binding proteins (PBPs). The product is intrinsically stable to dehydropeptidase-1 (DHP-1) degradation.
  • Active against susceptible and resistant *N. gonorrhoeae*, *H. influenzae*, and *H. ducreyi*.
  • Possesses broad-spectrum in vitro activity against many Gram-positive, Gram-negative, and anaerobic bacteria.
  • Significantly reduces the incidence of pancreatic infection in vivo.
Catalog No. 50-003-47955