Learn More
Medchemexpress LLC Meropenem trihydrate (SM 7338 trihydrate) | 119478-56-7 | 99.85% | 437.51 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY13678A500MG
Encompass_Preferred
Meropenem trihydrate is a carbapenem antibiotic with broad-spectrum antibacterial activity, inhibiting bacterial cell wall synthesis by binding to and inactivating penicillin-binding proteins (PBPs). It is effective against susceptible and resistant *N. gonorrhoeae*, *H. influenzae*, and *H. ducreyi*, and shows broad-spectrum in vitro activity against many Gram-positive, Gram-negative, and anaerobic bacteria.
  • Intrinsically stable to dehydropeptidase-1 (DHP-1) degradation
  • Effective against *N. gonorrhoeae*, *H. influenzae*, and *H. ducreyi*
  • Exhibits broad-spectrum in vitro activity
  • Significantly reduces the incidence of pancreatic infection in vivo
Catalog No. 50-003-48098