Learn More
ABclonal Technology MERS-CoV Nucleoprotein
SDP
Supplier:  ABclonal Technology A21179100UL
Encompass_Preferred
Packages the positive strand viral genome RNA into a helical ribonucleocapsid (RNP and plays a fundamental role during virion assembly through its interactions with the viral genome and membrane protein M Plays an important role in enhancing the efficiency of subgenomic viral RNA transcription as well as viral replication
Catalog No. 50-000-07851