Learn More
Sino Biological MERS-CoV (HCoV-EMC/2012) Spike Protein (Codon Optimized) Gene ORF cDNA clone expression plasmid, N-Flag tag
Supplier:  Sino Biological VG40069NF
Encompass_Preferred
Full length Clone DNA of MERS-CoV (NCoV / Novel coronavirus) Spike protein with N terminal Flag tag.
Catalog No. NC1794265