Learn More
Medchemexpress LLC Mertk protein, human (HEK293, His) | >95.0% | 50UG

Supplier: Medchemexpress LLC HYP70044250UG
Recombinant human MERTK protein (HEK293, His) is a C-terminally 10xHis-tagged receptor tyrosine kinase produced in HEK293 cells and supplied as a lyophilized powder. The protein corresponds to residues A21-I505 (UniProt Q12866), is glycosylated, and is suitable for biochemical assays, binding studies, and signaling research.
- Format: Lyophilized powder.
- Size: 50 μg.
- Purity greater than 95% by reducing SDS-PAGE.
- Predicted mass 55.4 kDa; observed ~120 kDa (glycosylated).
- Storage buffer: PBS with 6% trehalose, pH 7.4.
- Reconstitution: do not reconstitute below 100 μg/mL in ddH2O; aliquot and store at -20°C or -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.