Learn More
Bioworld MES Buffer, 1.0M, pH 5.5, 250 mL
Supplier:  Bioworld 413200862
Encompass_Preferred
Synonyms : 4-Morpholineethanesulfonic acidOne of the original Good’s buffers. MES solution has a pKa value near physiological pH, it is not absorbed through cell membranes, and it is essentially transparent to UV light. A zwitterionic buffer it is primarily used as a running buffer for resolving very small proteins on bis-tris gels. Also used in cation-exchange chromatography and as a biological buffer in plant cell cultures. MES solutions can be prepared to the desired pH by titrating the free acid with sodium hydroxide. The buffer solution once prepared should be stable at 2-8°C for months.Technical Information :pKa (25°C) : 6.10Warning: For laboratory use only.Not fit for agricultural, household, clinical use or human consumption.Appearance: LiquidMelting Point: >300 °C(lit.)PH Value: 5.5RTECS Number: QE3479500
Catalog No. NC1343333