Learn More
Medchemexpress LLC MES potassium (2-Morpholinoethanesulphonic acid potassium) | 39946-25-3 | 99.88% | 5 G
SDP
Supplier:  Medchemexpress LLC HYD0858E5G
Encompass_Preferred
MES potassium (2-Morpholinoethanesulphonic acid potassium) is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5-7.0. It is a Good's buffer widely used in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Amphoteric ion buffer with pH range 5.5-7.0.
  • Provides effective buffering capacity around pH 6.15.
  • pKa value changes with temperature, requiring consideration at varying temperatures.
  • Suitable for cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Exhibits poor affinity with metal ions, making it suitable for studies involving such ions.
Catalog No. 50-004-09963