Learn More
Alkali Scientific MES Sodium Salt [2 (N Morpholino)ethanesulfonic acid] sodium salt, 3 Kg
Supplier: Alkali Scientific ESS005CP
MES Sodium Salt [2-(N-Morpholino)ethanesulfonic acid] Sodium Salt, 3 Kg, is a powerful buffering agent used in various laboratory applications, particularly in the biological and chemical fields. This reagent helps maintain pH stability in experiments requiring a neutral to slightly acidic environment. MES is widely used in electrophoresis, enzyme assays, and cell culture media. With a capacity to buffer in the pH range of 5.5 to 6.7, it ensures reliable results in sensitive biological assays. The 3 kg package offers sufficient volume for ongoing experiments and provides an economical option for regular laboratory use in research and clinical applications.