Learn More
Research Products International Corp MES Sodium Salt [2-(N-Morpholino)ethanesulfonic acid] sodium salt, 500 Grams
SDP
Supplier:  Research Products International Corp M22050500.0
Encompass_Preferred
MES Sodium Salt, also known as 2-(N-Morpholino)ethanesulfonic acid sodium salt, is a zwitterionic buffer commonly used in biological and biochemical research.
In the life science field, MES Sodium Salt is primarily utilized as a buffering agent to maintain a stable pH in various experimental procedures. It is particularly effective in the pH range of 5.5 to 6.7, making it suitable for applications requiring slightly acidic to neutral pH conditions. 
MES Sodium Salt is commonly used in protein purification, enzyme assays, and electrophoresis techniques where a specific pH range needs to be maintained. Its buffering capacity and stability within the desired pH range make it valuable in these applications. 
One of the significant advantages of MES Sodium Salt is its compatibility with biological systems and enzymes. It exhibits minimal interference with enzymatic activity and other biological processes.
Catalog No. 50-997-915