Learn More
Apexbio Technology LLC MES sodium salt 71119-23-8 500mg
SDP
Supplier:  Apexbio Technology LLC C6532500
Encompass_Preferred
MES sodium salt (CAS 71119-23-8) is a buffering agent designed to maintain stable pH conditions in the range of 5 5 7 0 supporting physiological experimental environments MES sodium salt does not have a direct cellular or molecular target and is not a small-molecule inhibitor agonist or antagonist Its key role is pH stabilization in various biological and biochemical assays including enzymatic activity assays receptor pathway investigations and isolation of cellular organelles or biomolecules Experimental models include plant cell culture systems and physiological in vitro assays Based on these properties MES sodium salt holds research potential in applications such as plant cell culture controlled metabolic pathway studies and signal transduction research requiring consistent pH conditions
Catalog No. 50-000-92448