Learn More
Medchemexpress LLC Mesendogen | 864716-85-8 | 99.8% | 400.85 g·mol⁻¹ | C18H16ClF3N2OS | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10307325MG
Encompass_Preferred
Mesendogen is a small-molecule TRPM6 inhibitor used to promote mesoderm and definitive endoderm differentiation of human embryonic stem cells and induced pluripotent stem cells. It is used in research on magnesium homeostasis and TRPM6-related signaling during early embryonic development.
  • Promotes mesoderm and definitive endoderm differentiation.
  • Acts as a TRPM6 inhibitor for magnesium homeostasis studies.
  • High purity (99.84%) in a white to off-white solid form.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Suggested storage: powder -20°C for long-term stability.
Catalog No. 50-002-46037