Learn More
Strem, An Ascensus Company CAS# 917-23-7. 250mg. meso-Tetraphenylporphine TPP (chlorin free). MFCD00011680
SDP
Supplier:  Strem, An Ascensus Company 072170250MG
Encompass
CAS# 917-23-7. 250mg. meso-Tetraphenylporphine TPP (chlorin free). MFCD00011680. Molecular Weight: 614.75. Molecular Formula: C44H30N4. Color/form: violet xtl. Strem# 07-2170. http://www.strem.com/catalog/v/07-2170/
Catalog No. 50-901-09133