Learn More
Genetex Inc Mesothelin Monoclonal Antibody [MSVA-235M] HistoMAX™ (GTX04373) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04373
Encompass_Preferred
The Mesothelin antibody [MSVA-235M] HistoMAX™ (GTX04373) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human gastric cancer tissue using GTX04373 Mesothelin antibody [MSVA-235M] HistoMAX. Strong predominantly cytoplasmic mesothelin immunostaining in a stomach cancer with diffuse growth pattern.
Catalog No. NC3665620