Learn More
Genscript Corporation Mesothelin hFc Human
Supplier:  Genscript Corporation Z03636100
Encompass
Mesothelin also known as MSLN is a 40 kDa protein that is expressed in mesothelial cells It has been reported that this protein is over expressed in several human tumors including mesothelioma ovarian cancer pancreatic adenocarcinoma lung adenocarcinoma and cholangiocarcinoma Mesothelin can bind MUC16 (also known as CA125) indicating that the interaction of mesothelin and MUC16 may contribute to the implantation and peritoneal spread of tumors by cell adhesion Since mesothelin is overexpressed in several cancers and is immunogenic the protein could be exploited as tumor marker or as the antigenic target of a therapeutic cancer vaccine
Catalog No. 50-000-57143