Learn More
Medchemexpress LLC Mesothelin Human B 20ug
SDP
Supplier:  Medchemexpress LLC HYP7239820UG
Encompass_Preferred
Mesothelin is a glycosylphosphatidylinositol-anchored cell-surface protein that may function as a cell adhesion protein as cells overexpressing mesothelin have altered adhesive properties Mesothelin is also a Megakaryocyte-potentiating factor (MPF) which potentiates megakaryocyte colony formation in vitro Mesothelin may be related to cancer metastasis Mesothelin Protein Human (Biotinylated HEK293 His-Avi) is the recombinant human-derived Mesothelin protein expressed by HEK293 with C-Avi C-6 His labeled tag
Catalog No. 50-001-83081