Learn More
ABclonal Technology Mesothelin Rabbit mAb
SDP
Supplier:  ABclonal Technology A2379520UL
Encompass_Preferred
This gene encodes a preproprotein that is proteolytically processed to generate two protein products megakaryocyte potentiating factor and mesothelin Megakaryocyte potentiating factor functions as a cytokine that can stimulate colony formation of bone marrow megakaryocytes Mesothelin is a glycosylphosphatidylinositol-anchored cell-surface protein that may function as a cell adhesion protein This protein is overexpressed in epithelial mesotheliomas ovarian cancers and in specific squamous cell carcinomas Alternative splicing results in multiple transcript variants at least one of which encodes an isoform that is proteolytically processed
Catalog No. 50-000-06281