Learn More
Medchemexpress LLC Met-Arg-Phe-Ala | 67368-29-0 | 99.8% | 523.65 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP427010MG
Encompass_Preferred
Met-Arg-Phe-Ala is a peptide and a potent competitive inhibitor for enkephalin-generating endopeptidase (EGE). It can be used for the research of neurological disease.
  • Potent competitive inhibitor for enkephalin-generating endopeptidase (EGE).
  • Can be used for research of neurological disease.
  • White to off-white solid appearance.
  • Soluble in DMSO (≥ 100 mg/mL).
Catalog No. 50-003-97659