Learn More
Medchemexpress LLC MET kinase-IN-4 | 888719-03-7 | 99.0% | 458.42 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY18309
Encompass_Preferred
MET kinase-IN-4 is an orally active Met kinase inhibitor with potent Met kinase inhibitory activity (IC50 value of 1.9 nM). It can be used for cancer research. It also inhibits Flt-3 and VEGFR-2 kinases with IC50 values of 4 nM and 27 nM, respectively. MET kinase-IN-4 (3 μM) shows good metabolic stability in both human and mouse liver microsomes.
  • Orally active Met kinase inhibitor.
  • Potent Met kinase inhibitory activity with an IC50 value of 1.9 nM.
  • Inhibits Flt-3 and VEGFR-2 kinases with IC50 values of 4 nM and 27 nM, respectively.
  • Shows good metabolic stability in both human and mouse liver microsomes.
  • Possesses a favorable pharmacokinetic profile in mice.
  • Demonstrates significant in vivo antitumor activity in the GTL-16 human gastric carcinoma xenograft model.
Catalog No. 50-003-84109