Learn More
Abcam Met (pY1234/pY1235) + total Met ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1264511X96TESTS
Encompass_Preferred
Met (pY1234/pY1235) + total Met ELISA Kit is a Semi-quantitative ELISA kit for the measurement of Met (pY1234/pY1235) + total Met in Mouse, Rat, Human in Tissue Lysate, Cell Lysate samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2233468