Learn More
Microsolv Technology Corporation Metabolomics Development Kit, 4.6x150. 1 Cogent Diamond Hydride™ & 1 Cogent Bidentate C18™. 1 PK.
Supplier:  Microsolv Technology Corporation 4300015P2
Encompass_Preferred
This is an economical way to purchase two HPLC Columns that are often used with LCMS in Metabolomics and many other fields. Each Column has a 2.1mm Internal Diameter and is 150mm Long, packed with 4um particles and 100A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. Includes the Cogent Diamond Hydride™ and Cogent Bidentate C18™ columns.
Catalog No. 50-227-0853