Learn More
Lj Star Inc METACLAMP 4S
Supplier:  Lj Star Inc METACLAMP 4S
Encompass
METACLAMP SANITARY CLAMP DISC EA 624.00SIZE: 4" SANITARY CLAMP CONNECTIONRING MATERIAL: DUPLEX STAINLESS STEEL(1.4462)GLASS: BOROSILICATE TO DIN7080PER DRAWING NO: 180RATED: 150 PSIG @ 536° F
Catalog No. NC3793013