Learn More
Cayman Chemical metaFluoxetInhydrochLRId 10mg
SDP
Supplier:  Cayman Chemical 2215810 MG
Encompass_Preferred
An isomer of fluoxetine; has weaker SSRI activity than fluoxetine (Kis = 166 and 17 nM, respectively, in rat brain synaptosomes)
Catalog No. 50-298-7782