Learn More
Grainger Metal Hose 1-1/2 ID x 10 ft.L
Supplier:  Grainger 16T777
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 1" diameter x 25' long stainless steel metal hose with interlocking annular corrugations. It has a 300 PSI working pressure and is designed for use with air, water, oil, and other compatible fluids. The hose ends have 1" NPT male threaded fittings and it can handle temperatures from -40°F to 300°F. This durable, kink resistant metal hose is made in the USA.
Catalog No. 50-276-7247