Learn More
Grainger Metal Hose 2-1/2 ID x 10 ft.L
Supplier:  Grainger 16T835
Encompass_Preferred
The Hose Master Flexible Metal Hose Assembly is a stainless steel flexible metal hose with a 1" NPT male end and a 1" NPT female swivel end. It has a working pressure of up to 300 PSI and a maximum temperature of 850°F. The corrugated stainless steel hose allows flexibility while maintaining strength and durability. It has a length of 12" and is made in the USA. The flexible metal hose is ideal for connecting rigid piping that requires flexibility and vibration absorption. Common applications include engine exhaust, fume removal, and transferring liquids, air, and gases.
Catalog No. 50-276-8348