Learn More
Grainger Metal Hose 2 ID x 14 ft.L
Supplier:  Grainger 16T787
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 2" diameter, 25' long hose with interlocking segments for flexibility and durability. It has a working pressure of 300 PSI and a burst pressure of 1200 PSI. Made from 304 stainless steel with zinc-plated fittings, it operates from -40°F to 300°F. The design prevents kinking with a 4" minimum bend radius. It's used for hydraulic, lubrication, and transfer lines for petrochemicals, acids, caustics, gases, and more, offering flexibility and strength for industrial applications.
Catalog No. 50-276-7472