Learn More
Grainger Metal Hose 2 ID x 18 ft.L
Supplier:  Grainger 16T789
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 2" diameter, 25' long interlocked metal hose with a 300 PSI working pressure. It has zinc-plated steel fittings and is designed for use with air, water, oil and other non-corrosive fluids. This flexible metal hose allows easy maneuvering in tight spaces. It can handle temperatures from -40°F to 300°F. The interlocked construction provides strength while allowing flexibility. This industrial-grade hose is crush and kink resistant.
Catalog No. 50-276-7473