Learn More
Grainger Metal Hose 3 ID x 10 ft.L
Supplier:  Grainger 16T793
Encompass_Preferred
This stainless steel hose has an inside diameter of 3 inches and is available in lengths from 25 to 100 feet. It has a working pressure of up to 450 PSI and temperature range of -40 to 1000°F. The hose is covered in a 304 stainless steel braid for durability and has NPT threaded ends. It's designed for conveying liquids, gases, and solids in extreme environments. This heavy duty hose from Grainger is crush and kink resistant.
Catalog No. 50-276-5768