Learn More
Grainger Metal Hose 3 ID x 20 ft.L
Supplier:  Grainger 16T849
Encompass_Preferred
The Grainger 16T849 is a Westward 3/8-inch drive deep impact socket set. This 12-piece socket set includes sizes ranging from 4mm to 14mm, allowing you to work on a variety of fastener sizes. The sockets are made from chrome vanadium steel for durability and feature a black phosphate finish to resist corrosion. The deep design allows you to reach fasteners in tight spaces. This socket set comes organized in a molded case for convenient storage and portability. The Westward sockets meet ANSI standards and come with a lifetime guarantee against defects. This is a quality socket set for impact wrench use.
Catalog No. 50-276-8411