Learn More
Grainger Metal Hose 3 ID x 25 ft.L
Supplier:  Grainger 16T850
Encompass_Preferred
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or gas in industrial applications. It has a 304 stainless steel inner core and a 321 stainless steel outer braid. The hose has a 1/4 inch NPT male pipe thread on each end and is available in lengths from 12 inches to 20 feet. The working pressure is up to 1000 psi and the temperature range is -40°F to 1000°F. The metal hose is designed for durability and corrosion resistance.
Catalog No. 50-276-5887