Learn More
Grainger Metal Hose 4 ID x 10 ft.L
Supplier:  Grainger 16T802
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 4 inch diameter metal hose with a working pressure of 300 psi. It is made from 304 stainless steel with a smooth bore interior and is interlocked for flexibility. This metal hose has a temperature range of -20 to 650 degrees Fahrenheit. It has zinc-plated carbon steel fittings and is 30 feet in length. The hose is designed for conveying air, water, oil and other fluids in industrial applications.
Catalog No. 50-276-4963