Learn More
Grainger Metal Hose 4 ID x 10 ft.L
Supplier:  Grainger 16T853
Encompass_Preferred
The HOSE MASTER UFS18S0400 10AMM METAL HOSE 4" ID X 10' FTL 16T853 is a 4" diameter x 10' long stainless steel metal hose with a working pressure of 400 PSI. It has a 10 AMM end connection type and is made from 316 stainless steel for corrosion resistance. The hose allows for easy routing and installation while maintaining strength and durability. It can be used for hydraulic applications, air lines, water lines, chemicals, petroleum products, etc. The hose provides flexibility while handling high pressures.
Catalog No. 50-276-5888