Learn More
Grainger Metal Hose 4 ID x 12 ft.L
Supplier:  Grainger 16T854
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 4 inch diameter metal hose with a working pressure of 300 PSI. It has a stainless steel interlock construction and is designed for use with petroleum products, air, and water. The hose has a temperature range of -40°F to 300°F. It comes in a standard length of 25 feet and has male NPT threaded ends. The interlocked stainless steel design allows the hose to be flexible while still maintaining strength and durability. This metal hose would be well-suited for industrial applications requiring high working pressures.
Catalog No. 50-276-4884