Learn More
Grainger Metal Hose 4 ID x 16 ft.L
Supplier:  Grainger 16T856
Encompass_Preferred
The Grainger Approved Metal Hose is a flexible metal hose that connects air, water, oil, or exhaust systems. It has a stainless steel inner hose covered by a stainless steel wire braid exterior. The metal hose is available in sizes ranging from 1/4 inch to 2 inches in diameter, and lengths from 10 feet to 50 feet. It can withstand temperatures from -40°F to 1000°F. The metal hose is designed for automotive, industrial, and commercial applications that require a durable, high temperature and corrosion resistant flexible metal hose.
Catalog No. 50-276-6118