Learn More
Grainger Metal Hose 4 ID x 16 ft.L
Supplier:  Grainger 16T805
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 4-inch diameter metal hose with a working pressure of 300 PSI. It has interlocking annular corrugations for flexibility and is made from 304 stainless steel with a smooth inner wall for optimal flow. This hose meets SAE 100R2 standards and has a temperature range of -40°F to 1000°F. The fittings are zinc-plated steel fixed shank fittings. This durable metal hose is ideal for conveying air, water, oil and other fluids in industrial applications requiring high working pressures and temperatures.
Catalog No. 50-276-6646