Learn More
Grainger Metal Hose 5 ID x 10 ft.L
Supplier:  Grainger 16T810
Encompass_Preferred
The Hose Master Interlocked Metal Hose is a 5-inch diameter interlocked metal hose with a working pressure of 300 PSI. It has a stainless steel construction with a smooth bore interior and is designed for conveying air, water, oil and other fluids. This durable hose offers flexibility and can be used in a variety of industrial applications. The interlocked design allows it to be extended and compressed repeatedly without kinking. It connects using NPT threaded fittings. Overall, this is a heavy-duty, flexible metal hose for conveying fluids under pressure.
Catalog No. 50-276-6444